Learn More
Tocris Bioscience™ CP 376395 hydrochloride
Potent and selective CRF1 antagonist
Brand:  Tocris Bioscience™ 321210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13442574
  • £183.48 / 10mg
Estimated Shipment
08-07-2019
Description
Description
Potent and selective CRF1 receptor antagonist (Ki values are 12 and >10000 nM for CRF1 and CRF2 receptors respectively). Attenuates CRF-induced activation of the HPA axis in vivo following i.v. administration. Orally active.
Specifications
Specifications
CP 376395 hydrochloride
>99
10mg
CP 376395 hydrochloride
362.94