Learn More
Tocris Bioscience™ CP 376395 hydrochloride
Potent and selective CRF1 antagonist; binds at an allosteric site
POA
Specifications
Purity >99%
View More Specs
Description
Description
Potent and selective CRF1 receptor antagonist (Ki values are 12 and >10000 nM for CRF1 and CRF2 receptors respectively). Attenuates CRF-induced activation of the HPA axis in vivo following i.v. administration. Orally active.
Specifications
Specifications
>99%