Learn More
CP 339818 hydrochloride, Tocris Bioscience™
Non-peptide, potent KV1.3 channel blocker
Brand:  Tocris (Bio-Techne) 1399/50
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13459313
  • £582.37 / 50mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Potent, non-peptide KV1.3 channel antagonist that preferentially binds to the C-type inactivated state of the channel (IC50 ∽ 200 nM) . Inhibits KV1.4 with an IC50 of ∽ 300 nM. Selective over KV1.1, KV1.2, KV1.5, KV1.6, KV3.1-4, and KV4.2.
Specifications
Chemical Identifiers
478341-55-8
326.868
n-1-benzylquinolin-4-ylidene butan-1-amine hydrochloride
1-benzyl-N-butylquinolin-4-imine;hydrochloride
C20H23ClN2
HDAMNMJLKSDNJF-UHFFFAOYSA-N
71433722
CCCCN=C1C=CN(C2=CC=CC=C12)CC3=CC=CC=C3.Cl
Specifications
CP 339818 hydrochloride
478341-55-8
n-1-benzylquinolin-4-ylidene butan-1-amine hydrochloride
CCCCN=C1C=CN(C2=CC=CC=C12)CC3=CC=CC=C3.Cl
326.868
340.89
50mg
C20H23ClN2
HDAMNMJLKSDNJF-UHFFFAOYSA-N
1-benzyl-N-butylquinolin-4-imine;hydrochloride
71433722
>99%
Videos