missing translation for 'onlineSavingsMsg'
Learn More

CP 135807, Tocris Bioscience™

Selective 5-HT1D agonist

Brand:  Tocris (Bio-Techne) 378310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13443594

  • £204.33 / 10mg

Estimated Shipment date
from Supplier 23-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective 5-HT1D receptor agonist. Produces dose-dependent decreases in extracellular 5-HT concentration.
Specifications

Chemical Identifiers

151272-90-1
351.41
chembl83597, 3-n-methylpyrrolidin-2-ylmethyl-5-3-nitropyrid-2-ylamino-1h-indole, r-3-1-methyl-2-pyrrolidinyl methyl-n-3-nitro-2-pyridinyl-1h-indol-5-amine, 3-2r-1-methyl-2-pyrrolidinyl methyl-n-3-nitro-2-pyridinyl-1h-indol-5-amine, 1h-indol-5-amine, 3-1-methyl-2-pyrrolidinyl methyl-n-3-nitro-2-pyridinyl-, r, 3-2r-1-methylpyrrolidin-2-yl methyl-n-3-nitropyridin-2-yl-1h-indol-5-amine, cp hplc, 3-n-methylpyrrolidin-2r-ylmethyl-5-3-nitropyrid-2-yl amino-1h-indole
3-[[(2R)-1-methylpyrrolidin-2-yl]methyl]-N-(3-nitropyridin-2-yl)-1H-indol-5-amine
C19H21N5O2
YPFIYPNOWVPAPR-OAHLLOKOSA-N
5487088
CN1CCCC1CC2=CNC3=C2C=C(C=C3)NC4=C(C=CC=N4)[N+](=O)[O-]

Specifications

10mg
151272-90-1
chembl83597, 3-n-methylpyrrolidin-2-ylmethyl-5-3-nitropyrid-2-ylamino-1h-indole, r-3-1-methyl-2-pyrrolidinyl methyl-n-3-nitro-2-pyridinyl-1h-indol-5-amine, 3-2r-1-methyl-2-pyrrolidinyl methyl-n-3-nitro-2-pyridinyl-1h-indol-5-amine, 1h-indol-5-amine, 3-1-methyl-2-pyrrolidinyl methyl-n-3-nitro-2-pyridinyl-, r, 3-2r-1-methylpyrrolidin-2-yl methyl-n-3-nitropyridin-2-yl-1h-indol-5-amine, cp hplc, 3-n-methylpyrrolidin-2r-ylmethyl-5-3-nitropyrid-2-yl amino-1h-indole
CN1CCCC1CC2=CNC3=C2C=C(C=C3)NC4=C(C=CC=N4)[N+](=O)[O-]
351.41
351.4
CP 135807
C19H21N5O2
YPFIYPNOWVPAPR-OAHLLOKOSA-N
3-[[(2R)-1-methylpyrrolidin-2-yl]methyl]-N-(3-nitropyridin-2-yl)-1H-indol-5-amine
5487088
>98%
Videos
Documents