missing translation for 'onlineSavingsMsg'
Learn More

COX IV Mouse anti-Human, eFluor 570, Clone: IB52C31H10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 41-9775-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15508976

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The IB52C31H10 monoclonal antibody recognizes human COX IV. COX IV, also known as cytochrome c oxidase (COX) or Complex IV, is the terminal complex of the electron transport chain. COX IV is encoded by nuclear DNA and localized to the inner mitochondrial membrane. COX IV functions in energy metabolism and shuttles electrons from cytochrome c to oxygen. Dysfunction in COX IV is associated with increased production of reactive oxygen species and cellular toxicity. Genetic defects in COX IV results in severe, often fatal metabolic disorders that primarily affect tissues with high energy demands such as heart, brain, and muscle.
Specifications

Specifications

COX IV
IB52C31H10
eFluor 570
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Microscopy
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Cytochrome c oxidase, Subunit IV, Complex IV
IgG1, kappa
RUO
Primary
Human
Videos

For Research Use Only.