missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cover for General Purpose Water Bath

Fisherbrand™ Covers are for use withShallow-Form Shaking Water Baths.

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 1.14000kg

 View more versions of this product

Product Code. 15305897

  • £242.31 / Each


Estimated Shipment
31-05-2018
Add to basket

Specifications

Specifications

Precision 2L and 5L General Purpose Water Baths
Precision 2L and 5L General Purpose Water Baths
Gable Cover
Stainless Steel Gable Cover
Stainless Steel
Videos
Documentation