missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coumarin, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Coumarin is an anticoagulant found in medicinal plants and used in phytomedicine for the treatment of venous diseases. This compound is found to have anti-tumor activities in several human cells lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 68°C to 71°C |
Density | 0.935 |
Boiling Point | 297°C to 299°C |
Flash Point | 162°C (323°F) |
Odor | Pleasant |
Quantity | 250 g |
UN Number | UN2811 |
Beilstein | 383644 |
Merck Index | 14,2562 |
Solubility Information | Soluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'