missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 343, Laser Grade, Thermo Scientific Chemicals

Catalog Number p-94000
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
55804-65-4
C16H15NO4
285.299
MFCD00051335
KCDCNGXPPGQERR-UHFFFAOYSA-N
108770
CHEBI:51941
C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C(=O)O)CCCN3C1
This item is not returnable. View return policy
55804-65-4
C16H15NO4
285.299
MFCD00051335
KCDCNGXPPGQERR-UHFFFAOYSA-N
108770
CHEBI:51941
C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C(=O)O)CCCN3C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 240.0°C
Fluorescence Max 470nm (in basic ethanol)
Solubility Information Solubility in water: slightly soluble.
Infrared Spectrum Authentic
Lambda Max 409nm (in basic ethanol)
Formula Weight 285.29
Grade Pure
Molar Absorption Coefficient 19900L/(mole.cm) (lambda max.)
Packaging Glass bottle
Suitability (for use as a laser dye) passes
Color Orange
Quantity 100 mg
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 343
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'