missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 314, 99%, pure, laser grade, Thermo Scientific Chemicals

Catalog Number p-4837995
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
This item is not returnable. View return policy
55804-66-5
C18H19NO4
313.353
MFCD00051334
VMJKUPWQKZFFCX-UHFFFAOYSA-N
1,2,4,5,3H,6H,10H-Tetrahydro-9-carbethoxy[1]benzopyrano(9,9a,1-gh)-quinolizin-10-one
72653
CHEBI:51940
CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3
This item is not returnable. View return policy
55804-66-5
C18H19NO4
313.353
MFCD00051334
VMJKUPWQKZFFCX-UHFFFAOYSA-N
1,2,4,5,3H,6H,10H-Tetrahydro-9-carbethoxy[1]benzopyrano(9,9a,1-gh)-quinolizin-10-one
72653
CHEBI:51940
CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 140.0°C to 144.0°C
Assay Percent Range 99%
Formula Weight 313.34
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Quantity 100 mg
Chemical Name or Material Coumarin 314
compliance-icons
Product Identifier
  • Coumarin 314, laser grade
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'