missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog Number 10695951
Click to view available options
Color:
Clear
No. per Pack:
1/Pk.
10/Pk.
20/Pk.
5/Pk.
No. per Case:
100/Cs.
50/Cs.

Color: Clear; Sterility: Sterile; Lid: With Lid

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

TRUSTED_SUSTAINABILITY
Format Standard
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Binding Type TC-Treated
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 0.1 to 0.2 mL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 0.36 mL
Well Shape Flat Bottom (F-well)
No. per Case 50/Cs.
No. per Pack 1/Pk.
Quantity 50/Cs.
Well Color Clear
Diameter (Metric) Well 0.64 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'