Learn More
Corning™ Costar™ Flat Bottom Cell Culture Plates
Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.
£122.55 - £352.84
Specifications
Color Clear
Sterility Sterile
Lid With Lid
Material Polystyrene
View More Specs
Products 13
Product Code Brand Format Culture Area No. per Case Lid Price Quantity & Availability  
Product Code Brand Format Culture Area No. per Case Lid Price Quantity & Availability  
10792552 Corning™
3599
96 well 0.32cm2 100 (Individually wrapped) With Lid £280.53
Pack of 100 View alternative products
Save Up to 
»
In Stock 35
Add to basket
10065370 Corning™
3548
48 well 0.95cm2 100 (Individually wrapped) With Lid £352.84
Pack of 100 View alternative products
Save Up to 
»
In Stock 10
Add to basket
10380932 Corning™
3526
24 well 1.9cm2 50 (Individually wrapped) With Lid £128.16
Pack of 50 View alternative products
Save Up to 
»
In Stock 43
Add to basket
10377841 Corning™
3527
24 well 1.9cm2 100 (20 packs) With Lid £250.54
Pack of 100 View alternative products
Save Up to 
»
In Stock 11
Add to basket
10146810 Corning™
3506
6 well 9.5cm2 100 (20 packs) With Lid £242.17
Pack of 100 View alternative products
Save Up to 
»
In Stock 37
Add to basket
10136810 Corning™
3512
12 well 3.8cm2 100 (20 packs) With Lid £275.59
Pack of 100 View alternative products
Save Up to 
»
In Stock 6
Add to basket
10253041 Corning™
3513
12 well 4cm2 50 (Individually wrapped) With Lid £160.20
Pack of 50 View alternative products
Save Up to 
»
In Stock 53
Add to basket
10578911 Corning™
3516
6 well 9.5cm2 50 (Individually wrapped) With Lid £122.55
Pack of 50 View alternative products
Save Up to 
»
In Stock 175
Add to basket
10732552 Corning™
3524
24 well 1.9cm2 100 (Individually wrapped) With Lid £256.19
Pack of 100 View alternative products
Save Up to 
»
In Stock 42
Add to basket
10298901 Corning™
3585
96 well 0.32cm2 50 (10 packs) With Lid £151.92
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment
17-05-2019
Add to basket
10687551 Corning™
3595
96 well with low-evaporation lid 0.32cm2 50 (Individually wrapped) With low-evaporation lid £144.40
Pack of 50 View alternative products
Save Up to 
»
In Stock 79
Add to basket
10412741 Corning™
3598
96 well 0.32cm2 100 (20 packs) With Lid £260.41
Pack of 100 View alternative products
Save Up to 
»
In Stock 30
Add to basket
10695951 Corning™
3596
96 well 0.32cm2 50 (Individually wrapped) With Lid £141.06
Pack of 50 View alternative products
Save Up to 
»
In Stock 41
Add to basket
Description
Description
  • Designed for a wide range of applications, including general cell growth experiments, cloning studies, virus isolation and susceptibility testing, and in vitro testing
  • Plate bottoms feature uniform thickness—allowing for distortion-free observation and microscopic examination
  • Alphanumeric coordinates placed on the same focal plane as cell growth—for convenient referencing of well position
  • Gripping edges make handling easier
  • All plates have nonreversible lids with condensation rings to minimize evaporation and risk of contamination
  • Made of optically clear, medical-grade polystyrene 
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic
Certifications
Certified nonpyrogenic
Specifications
Specifications
Clear
Sterile
With Lid
Polystyrene