missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
GREENER_CHOICE

Catalog Number 10360691
Click to view available options
Packaging:
Bag of 20; Cs. of 100
Individually wrapped, 50/Cs.

Color: Clear; Sterility: Sterile; Lid: With Lid

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile
TRUSTED_SUSTAINABILITY
Format Standard
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Packaging Individually wrapped, 50/Cs.
Binding Type TC-Treated
Color Clear
Product Type Cell Culture Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 0.33 mL
Well Shape Round Bottom (U-well)
For Use With (Application) Attachment and growth of anchorage-dependent cells
No. per Case 50/Cs.
No. per Pack 1/Pk.
Quantity 50/Cs.
Well Color Clear
Diameter (Metric) Well 0.64 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'