Learn More
Corning™ Clear Polystyrene 96-Well Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
Brand:  Corning™ 3797
Code : NEW
Additional Details : Weight : 5.89670kg
 View more versions of this product
Product Code. 11942151
  • £560.00 / Pack of 960
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Medical-grade polystyrene
  • Well volume: 320µL to 360µL
  • Working volume: 75µL to 200µL
  • Choice of flat, round, flat Easy Wash™ or V-shaped bottom
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
  • Plate dimensions meet proposed industry standards
Specifications
Specifications
EIA/RIA
Round
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
4 x 25
330μL
Without Lid
Medium Binding
Clear
Polystyrene
Non-sterile
75 to 200μL
Videos