Learn More
Corning™ 96-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Brand: Corning™ 3922
Code : 66
Additional Details : Weight : 6.98532kg
Description
Specifications
Standard | |
8 x 12 | |
White | |
Polystyrene | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
Without Lid | |
4 inner packs of 25 | |
EIA/RIA | |
360 μL | |
75 to 200 μL | |
High Binding | |
Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions. | |
25/Pk. |