missing translation for 'onlineSavingsMsg'
Learn More

Cortisone, 98%, Acros Organics™

£13.30 - £26.18

Chemical Identifiers

CAS 53-06-5
Molecular Formula C21H28O5
Molecular Weight (g/mol) 360.44
InChI Key MFYSYFVPBJMHGN-ZPOLXVRWSA-N
Synonym cortisone, andreson, cortisal, cortisate, cortistal, cortivite, cortogen, cortone, kendall's compound e, adrenalex
PubChem CID 222786
ChEBI CHEBI:16962
IUPAC Name (8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione
SMILES CC12CCC(=O)C=C1CCC3C2C(=O)CC4(C3CCC4(C(=O)CO)O)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15559523 View Documents Acros Organics
460522500
250mg £13.30
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 18-06-2020
Please to purchase this item. Need a web account? Register with us today!
15549523 View Documents Acros Organics
460520010
1g £26.18
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-06-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

53-06-5
360.44
cortisone, andreson, cortisal, cortisate, cortistal, cortivite, cortogen, cortone, kendall's compound e, adrenalex
CHEBI:16962
CC12CCC(=O)C=C1CCC3C2C(=O)CC4(C3CCC4(C(=O)CO)O)C
C21H28O5
MFYSYFVPBJMHGN-ZPOLXVRWSA-N
222786
(8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione

Specifications

Cortisone
2% max.
250mg
97.5% min. (HPLC)
cortisone, andreson, cortisal, cortisate, cortistal, cortivite, cortogen, cortone, kendall's compound e, adrenalex
CC12CCC(=O)C=C1CCC3C2C(=O)CC4(C3CCC4(C(=O)CO)O)C
360.44
CHEBI:16962
98%
Authentic
223.0°C to 228.0°C
53-06-5
C21H28O5
MFYSYFVPBJMHGN-ZPOLXVRWSA-N
(8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione
222786
360.44
Safety and Handling

Safety and Handling

Product Identifier
  • Cortisone

Signal Word
  • Warning

Hazard Category
  • Reproductive toxicity Category 2

Hazard Statement
  • H361d-Suspected of damaging the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.