Learn More
Corning Biocoat™ Corning™ transforming growth factor-b (TGF-b), human natural, 50μg
Brand: Corning Biocoat™ 356040
Code : 67
Additional Details : Weight : 0.00800kg Transport : UN number : 1648 Chem class : 3 Pack group : II
Description
Corning Transforming Growth Factor-beta (TGF-beta), human natural, is a multifunctional protein that plays a central role in the regulation of cell growth and differentiation with either stimulatory or inhibitory effects.- Formulation: frozen in 36% acetonitrile-0.1% trifluoroacetic acid (caution - TGF-beta absorbs to glass and plastic surfaces)
- Sourced from human platelets. NOTE: source material tested for hepatitis B antigen and HIV-1 antibody
- Molecular weight 25kD
- Purity more than 95% by SDS-PAGE
- Tested for ability to inhibit DNA synthesis of Mv 1 Lu mink lung cells (ATCC CCL-64)
- Tested and found negative for bacteria, fungi and mycoplasma
- Recommended concentration 10-50 pg/mL of medium depending on cell type and application
- Stable for at least three months at -70°C (if diluted according to instructions)
Specifications
Frozen | |
36% acetonitrile-0.1% trifluoroacetic acid | |
>95% by SDS-PAGE |