missing translation for 'onlineSavingsMsg'
Learn More

Corning™ T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Rat

Promotes polyclonal activation and IL-2 proliferation of most antigenically sensitized rat or mouse lymphoid T-cells

Brand:  Corning™ 354116

Code : 67

Additional Details : Weight : 0.15000kg

 View more versions of this product

Product Code. 11523540

  • £782.67 / 100mL


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Description

Description

 

  • Conditioned medium (RPMI 1640) from ConA-stimulated rat splenocyte cultures containing 10% fetal bovine serum and 10mM HEPES
  • Medium with ConA supports high-titer HIV production by leukocytes
  • Medium without ConA (from washed rat splenocyte cultures) designed for applications that support proliferation of lectin-activated T-cells and single-cell cloning of antigen-specific T-cells from polyclonally activated primary cultures
  • Both undergo an appropriate bioassay, are filtered through a 0.2μm membrane and are tested for the absence of bacteria, fungi and Mycoplasma—guaranteeing a high-quality product

 

Specifications

Specifications

≤0.5μg/mL
T-cell culture Supplement, Without ConA
Rat
ConA
Max ConA Content: ≤ 0.5μg/mL
100mL
2° to 8°C
Videos