Learn More
Corning™ Surface Modified Stripwell™ Microplates
Four surface chemistries suit every type of assay
Brand:  Corning™ 2508
Code : 67
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10499131
  • £587.14 / Pack of 50
Estimated Shipment
23-02-2018
Add to basket
Specifications
Specifications
Microplate Strip
In-vitro diagnostic assays
Polystyrene
Non-sterile
75 to 200μL
Round
Clear
96 well
96
Carbo-Bind™
Without Lid
360μL
Videos
Documentation
Technical Documents