Learn More
Stripwell™ Microplates
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.
Brand:  Corning 2484
Code : 67
Additional Details : Weight : 6.80388kg
 View more versions of this product
Product Code. 10215362
  • £630.59 / Pack of 100
Estimated Shipment
30-07-2018
Add to basket
Description
Description
  • Some units contain twelve 1 × 8 strips pre-assembled in an “egg crate” style holder
  • Strips fit in frame in only one direction, eliminating the chance of misorientation
  • Flat-bottom wells can be easily snapped apart to allow for radiometric analysis
  • Hydrophobic medium-binding surface has a binding capacity of ∽250ng IgG/cm2 and is suited for large hydrophobic biomolecules Hydrophobic and ionic (negatively charged) high-binding surface has a binding capacity of ∽500ng IgG/cm2 and is ideal for use with positively charged biomolecules (>10kDa)
  • Available in standard volume or low volume to save antibody and reagent costs
  • Nonsterile
Specifications
Specifications
Microplate Strip
White
96 well
96
Non-sterile
Without Lid
190μL
Medium Binding
In-vitro diagnostic assays
Polystyrene
100/Cs.
75 to 125μL
Round
Videos
Documentation
Technical Documents