Learn More
Corning™ Storage Mat Applicator
Quickly applies Storage Mat III to 96-well plates
Brand: Corning™ 3081
Code : 66
Additional Details : Weight : 5.76062kg
Description
Storage Mat Applicators evenly apply storage mats to Corning 96 well polypropylene plates, blocks, and 96 well polystyrene plates.Specifications
Storage Mat Applicator | |
Black | |
For high-volume sample processing | |
1 Ea. |