Learn More
Corning™ Smooth 1536-Well Multiple Well Plates
Ensure precision liquid handling
Brand:  Corning™ 7247
Code : 66
Additional Details : Weight : 3.24000kg
 View more versions of this product
Product Code. 12761935
  • £835.86 / Pack of 50
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
Description
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Specifications
Specifications
Standard
32 x 48
1536-Well Assay Microplate
Polystyrene
1536
Non-sterile
Flat Bottom (F-well)
50/Cs.
Without Lid
White
Automated dispensers including the PerkinElmer FlexDrop™
12.5 μL
8 μL
Non-treated
For Kalypsys™ systems, ensure precision liquid handling
10/Pk.