Learn More
Corning™ SF Medium
Based on a proprietary combination of classical media formulations
Brand:  Corning™ 40-102-CV
Code : NEW
Additional Details : Weight : 3.76000kg
 View more versions of this product
Product Code. 15333701
  • £138.48 / Pack of 6
Estimated Shipment date
from Supplier 29-06-2018
Add to basket
Description
Description
  • Contains a mixture of select trace elements and high molecular carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1.0 g/L)
  • Supports superior growth and viability over long-term passages in hybridomas, as well as suspension and adherent cell cultures
  • Formulation contains L-glutamine
Offers a viable alternative to FBS for many hybridomas and works on a broad range of suspension and attachment cells; can be cell line dependent
Specifications
Specifications
7.2 to 7.6
6 x 500mL
2° to 8°C
SF Medium, With L-Glutamine; without Phenol Red
Yes
Phenol Red
Videos