Learn More
Corning™ Robolids with Corner Notch
For use with automation equipment
Brand:  Corning™ 3090
Code : 66
Additional Details : Weight : 2.85763kg
 View more versions of this product
Product Code. 10702783
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Combines the sealing ability of a storage mat with the rigidity of a plastic lid
  • Designed for repeated application and removal by automation and for preventing short-term evaporation
  • Silicone sealing plugs for organic solvent resistance and low extractables
  • Can be used manually or with automation
  • Order Info
    Available in 96- and 384-well formats.
    Specifications
    Specifications
    Non-sterile
    50/Cs.
    96-Well Robolid
    96
    25/Pk.