Learn More
Corning™ PureCoat™ Fibronectin Peptide Cell Culture Flasks with Vented Cap
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
£491.00 - £585.00
Specifications
Material | Polystyrene |
---|---|
Neck Style | Canted |
No. per Case | 10/Cs. |
No. per Pack | 5/Pk. |
Shape | Rectangular |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15350001
|
Corning™
356242 |
75 cm2 |
£491.00
Pack of 10 |
Estimated Shipment: 12-04-2024
Log in to see stock. |
|||||
15380001
|
Corning™
356243 |
175 cm2 |
£585.00
Pack of 10 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Specifications
Polystyrene | |
10/Cs. | |
Rectangular | |
Vented |
Canted | |
5/Pk. | |
PureCoat E cm Mimetic Fibronectin Peptid |