Learn More
Corning™ PureCoat™ Collagen Peptide Cell Culture Flasks with Vented Cap
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
Brand: Corning™ 356273
Code : 67
Additional Details : Weight : 2.02000kg
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Specifications
175 cm2 | |
Canted | |
5/Pk. | |
PureCoat E cm Mimetic Collagen I Peptide | |
Vented |
Polystyrene | |
10/Cs. | |
Rectangular | |
Rectangular | |
175 cm2 |