Learn More
Corning™ PureCoat™ Carboxyl Rectangular Canted Neck Cell Culture Flasks with Vented Cap
Negatively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays
Brand: Corning™ 354780
Code : 66
Additional Details : Weight : 0.95000kg
Description
Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.Specifications
25.3 oz. | |
Polystyrene | |
Canted | |
5 Pk. | |
Carboxyl |
750 mL | |
For preparative cell culture, cell-based and HTS screening assays | |
5/Cs. | |
Rectangular | |
Vented |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.