Learn More
Corning™ Primaria™ Tissue Culture Flasks
Improves attachment, spreading and growth for many primary cells or cell lines. Corning™ Primaria™ Tissue Culture Flasks are suitable for growing cells that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes.
Brand:  Corning™ 353808
Code : 66
Additional Details : Weight : 5.21630kg
 View more versions of this product
Product Code. 10613672
  • £198.25 / Pack of 100
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
  • Unique nitrogen-containing tissue culture surface chemistry
  • Improves attachment, spreading, and growth for many primary cells or cell lines
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
Specifications
Specifications
Blue
50 mL
Canted
20/Pk.
Sterile
Vented
Phenolic
Polystyrene
100/Cs.
Rectangular
Rectangular
25 cm2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.