missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 2.5S Nerve Growth Factor (NGF), Mouse Natural

Designed specifically for use in basic cell biology research

Brand:  Corning™ 356004

Code : 66

Additional Details : Weight : 0.02350kg

 View more versions of this product

Product Code. 11503610

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance

Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Description

Description

  • NGFs are isolated from mouse submaxillary glands
  • 2.5S form consists of two β subunits
  • 7S form consists of two α, one β and two ϒ subunits
  • Products are tested for biological activity in appropriate assays, such as the ability of growth factors to stimulate proliferation or DNA synthesis
  • Lymphokines and growth factors are also sterile filtered generally using a 0.2μm membrane
  • Tested for the presence of bacteria, fungi and mycoplasma

Specification sheets include detailed information on quality control testing, product source, storage, stability and recommendations on proper experimental concentrations.

Order Info

Both supplied in lyophilized form

Specifications

Specifications

Mouse
Lyophilized from phosphate buffered saline, carrier free, reconstitute in serum containing medium or aqueous buffer containing BSA 2%
26
100μg
2° to 8°C
Nerve Growth Factor
Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
Yes
>90% by 4 to 20% Tris Glycial Gel Electrophoresis
3 Months
Mouse Submaxillary Glands
Videos