Corning™ Interleukin-2 (IL-2), Mouse Recombinant

Elicit a range of responses from many cell types

Overview
Brand: Corning™

Manufacturer Part Number: 354078

25000UNIT BRMP Interleukin-2 (IL-2), Mouse Reco

UNSPSC: 12352205

Code: 67

Additional Details:
Additional Details: Weight: 0.02650kg



Alerts: In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In early January 2014, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Product Code. 10113721

Quantity Price
1 £ 404.25 / 25000 units
Estimated Shipment date
from Supplier 27-02-2017
Add to basket


Description and Specification

Specification

Formulation Frozen in Dulbecco's PBS containing 0.1% BSA
Purity >95% by SDS-PAGE analysis
Quantity 25000U
Shelf Life 3 Months
Source Recombinant murine T-cell line
Sterile Yes
Storage Requirements -70°C
Product Type IL-2, mouse recombinant