Learn More
Corning™ Microvolume Pipet Tips, Nonsterile
Provide accurate, reliable performance in the stated volume range
Brand: Corning™ 4840
Code : 66
Additional Details : Weight : 1.67829kg
Description
- Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
0.2 to 10 μL | |
Natural | |
Bulk | |
Bulk | |
Polypropylene |
Non-sterile | |
DNase, RNase, and Pyrogen free | |
Microvolume, Round | |
Gilson™ Pipetman™, Universal Fit Ultramicro |