Learn More
Corning™ Disposable Glass Pasteur Pipets, Non-Sterile, Unplugged
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
Brand: Corning™ 7095B-NMR
Code : 66
Additional Details : Weight : 0.38700kg
Description
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
Corning disposable glass Pasteur pipets are available in convenient Packages. One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents. These pipets feature a constriction one inch below the top to facilitate plugging. They are ideal for rapid nonvolumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.
'B' series pipets are soda lime glass. 'D' series are borosilicate glass.
Specifications
No | |
0.65 cm | |
1 in. Constriction below the top | |
Round | |
Unplugged Pasteur Pipet |
0.255 in. | |
Soda Lime Glass | |
13 in. | |
33.02 cm | |
Single-use |