missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DeckWorks™ Tip Stations, Nonsterile

Designed for single and multichannel pipetting stations

Brand:  Corning™ 4142

Code : A0

Additional Details : Weight : 3.45000kg

 View more versions of this product

Product Code. 11901861

  • £48.74 / Pack of 960


Estimated Shipment
01-03-2018
Add to basket

Description

Description

  • Contains 960 tips available in extended length 10μL and traditional 200μL sizes
  • Maximizes bench space while minimizing plastic waste
  • The 5-layer deck design (2 × 96) matches the microtiter plate format
  • This stable pipetting platform fits easily into sterile hood environments
  • Lids can be snap-positioned to expose a single side (96 tips) only
  • Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities
  • Either sterile or nonsterile

Specifications

Specifications

Tip Station
Nonsterile
Yes
Polypropylene
1 to 200μL
Natural
Round
Multipack
Videos