missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DeckWorks™ Tip Stations, Nonsterile

Designed for single and multichannel pipetting stations

Brand:  Corning™ 4141

Code : 67

Additional Details : Weight : 3.40194kg

 View more versions of this product

Product Code. 11363665

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Contains 960 tips available in extended length 10μL and traditional 200μL sizes
  • Maximizes bench space while minimizing plastic waste
  • The 5-layer deck design (2 × 96) matches the microtiter plate format
  • This stable pipetting platform fits easily into sterile hood environments
  • Lids can be snap-positioned to expose a single side (96 tips) only
  • Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities
  • Either sterile or nonsterile

Specifications

Specifications

Multipack
Nonsterile
Yes
PP (Polypropylene)
0.1 to 10μL
Natural
Round
Videos