Learn More
Corning™ DeckWorks Pipet Tips, Nonsterile
Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking
Brand:  Corning™ 4115
Code : 67
Additional Details : Weight : 6.48637kg
 View more versions of this product
Product Code. 11941841
  • £58.75 / Pack of 960
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Available in 10μL, 200μL, 300μL and 1000μL sizes
  • The 10μL, 300μL and 1000μL tips are extended length and allow improved access into micro (1.5mL) and standard conical bottom (15/50mL) centrifuge tubes
  • Robust 96-tip racks are designed for repeated steam autoclave cycles
  • Can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system
  • Allow maximize storage space and help reduce the production and waste of plastic materials
  • Hinged racks within this system are manufactured from recycled polypropylene
  • Corning racked 300μL DeckWorks are designed for popular 300μL multi-channel pipettors
Specifications
Specifications
Hinged Rack
Universal Fit
Nonsterile
Round
Hinged Rack
0.1 to 10μL
Natural
Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking
Videos