missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ DeckWorks™ Low Binding Hinged Rack Pipette Tips
Used for quantitative assays in which sample binding can significantly alter results.
Brand: Corning™ 4139
Code : 67
Additional Details : Weight : 1.63350kg
Description
- Low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures.
- Low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.
Specifications
1 to 200 μL | |
Sterile | |
Used for quantitative assays in which sample binding can significantly alter results | |
Low Retention, Round | |
Yes | |
HDPE |
Filtered | |
Natural | |
Hinged Rack | |
Rack | |
DeckWorks |