Learn More
Corning™ DeckWorks™ Low Binding Hinged Rack Pipette Tips
Used for quantitative assays in which sample binding can significantly alter results.
Brand:  Corning™ 4139
Code : 67
Additional Details : Weight : 8.25537kg
Product Code. 11343665
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • The hinged rack design allows easy rack stacking
  • Racks can be reused
  • Simply remove the tip insert in the rack and replace with the Corning tip inserts for hinged racks
  • Proprietary low binding technology reduces surface tension on the interior wall of the tip
  • Allow maximize storage space and help reduce the production and waste of plastic materials
  • Hinged racks within this system are manufactured from recycled polypropylene
  • These sterile tips will not trap liquids or inhibit PCR in the event of over-pipetting
Model No. 10-320-728: The unique design of this extended-length tip allows standardization across wide range of 20mL, 50mL, 100mL and 200mL pipettors.
Specifications
Specifications
Low Binding Pipette Tip
Sterile
Yes
Round
Hinged Rack
Used for quantitative assays in which sample binding can significantly alter results
1 to 200μL
Natural
Nonfilter, Hinged Rack
HDPE
768 Tips/Pack, 4 Pack/Case
Videos