Learn More
Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips
Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures
Brand:  Corning™ 4138
Code : 67
Additional Details : Weight : 7.25747kg
 View more versions of this product
Product Code. 11333665
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Corning DeckWorks low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.
Specifications
Specifications
1 to 200 μL
Sterile
Used for quantitative assays in which sample binding can significantly alter results
Low Retention, Round
Yes
HDPE
Filtered
Natural
Hinged Rack
Rack
DeckWorks