Learn More
Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips
Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures
Brand: Corning™ 4136
Code : 67
Additional Details : Weight : 6.71316kg
Description
Corning DeckWorks low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.
Specifications
1 to 20 μL | |
Sterile | |
Used for quantitative assays in which sample binding can significantly alter results | |
Low Retention, Round | |
Yes | |
HDPE |
Filtered | |
Natural | |
Hinged Rack | |
Rack | |
DeckWorks |