Learn More
Corning™ General Assay Microplate Lids
Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.
Brand:  Corning™ 3930
Code : A0
Additional Details : Weight : 0.12972kg
 View more versions of this product
Product Code. 12647795
  • £14.15 / 10m
In Stock 8
Add to basket
You may only order up to the available stock. For more details call customer services on 01509 555 500
Description
Description
  • All lids are made of rigid polystyrene 
  • Lids with a corner notch on the A1 corner correspond to the corner notches found on all Corning® microplates 
  • The Universal Lid (without a corner notch) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids
Specifications
Specifications
Sterile
Low evaporation lid with corner notch and condensation rings
96 well microplates only (not 2mL block)
1.0
Polystyrene
Lid w/ corner notch
100
Videos