missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ Sodium Bicarbonate, Powder

Part of buffering system commonly used to maintain physiological pH 7.2 to 7.4 in culture environment

Brand:  Corning™ 61-065-RR

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15383751

  • £165.10 / 1000g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • Molecular weight: 84.01

Specifications

Specifications

Powder
6.8 to 7.4
Sodium Bicarbonate
15° to 30°C
Plastic Bottle
1kg
Sterile-filtered
Videos