missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Microbiological Media
Sterile filtered
Brand: Corning™ 46-050-CM
Code : NEW
Additional Details : Weight : 6.98000kg
Description
- For the maintenance and propagation of E. coli
- Terrific Broth supports higher cell densities than LB Broth, increasing plasmid yield
Specifications
Liquid | |
LB Broth | |
Casein Peptone 10g/L, NaCl 10g/L | |
Bottle |
United States | |
6 x 1000 mL | |
For maintenance/propagation of Escherichia coli with plasmid DNA and recombinant Protein production | |
6 x 1000 mL |