missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ L-glutamine Powder

An essential amino acid and a key component of culture media

Brand:  Corning™ 61-030-RM

Code : NEW

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 15323751

  • £95.27 / 100g


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • Serves as major energy source for propagating cells
  • Molecular Weight: 146.15

Specifications

Specifications

Powder
5 to 6
L-Glutamine
Plastic Bottle
100g
2° to 8°C
Videos