missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ HEPES, Powder

An alternative to bicarbonate for buffering

Brand:  Corning™ 61-034-RM

Code : NEW

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 15353751

  • £104.80 / 100g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Description

Description

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • pH 7.2 to 7.6
  • Molecular Weight: 238.3

Specifications

Specifications

Powder
6.8 to 8.2
HEPES
15° to 30°C
Plastic Bottle
100g
Sterile-filtered
Videos