Corning™ cellgro™ Gentamicin Sulfate

Packaged in plastic bottles

Overview
Brand: Corning™

Manufacturer Part Number: 61-098-RF

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: CAS Number: 1405-41-0 Weight: 0.01000kg



Product Code. 15313761

Quantity Price
1 £ 62.4 / 1g
Estimated Shipment date
from Supplier 03-03-2017
Add to basket


Description and Specification

Specification

Storage 2° to 8°C
Storage Requirements 2° to 8°C
Solution Type Gentamicin Sulfate
Sterile Yes
Format Powder
Format Powder
Packaging Plastic Bottle
Quantity 1 x 1g
Packaging Type Plastic Bottle
Quantity 1 x 1g
Sterile Yes