missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ Gentamicin Sulfate

Packaged in plastic bottles

Brand:  Corning™ 61-098-RA

Code : NEW

Additional Details : CAS Number : 1405-41-0 Weight : 0.01000kg

 View more versions of this product

Product Code. 15303761

  • £44.06 / 0.10g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Specifications

Powder
1 x 100mg
Yes
2° to 8°C
Plastic Bottle
Gentamicin Sulfate
2° to 8°C
Videos