missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ G418 Sulfate, Powder

Packaged in plastic bottles

Brand:  Corning™ 61-234-RF

Code : NEW

Additional Details : CAS Number : 108321-42-2 Weight : 0.01000kg

 View more versions of this product

Product Code. 15303771

  • £118.46 / 1g


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Specifications

Specifications

>700mcg/mL
Powder
G418 Sulfate, Powder
2° to 8°C
CAS: 108321-42-2
1g
2° to 8°C
Videos