missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DMEM/Ham's F-12 50/50 Mix, Powder

Used for culture of variety of cell types under low serum conditions. Corning™ DMEM/Ham's F-12 50/50 Mix, Powder ensures improved stability.

Brand:  Corning™ 90-091-PBR

Code : NEW

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 15393781

  • £35.29 / 10 L


Estimated Shipment date
from Supplier 24-07-2018
Add to basket

Description

Description

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • Formulation is without sodium bicarbonate, L-glutamine and phenol red
Specifications

Specifications

1X
DMEM/Ham's F-12 50/50 Mix
2° to 8°C
Powder
1 x 10L
Sodium Bicarbonate and L-Glutamine
Videos