missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CellBIND™ Surface Cell Culture Flasks

Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.

Brand:  Corning™ 3292A

Code : NEW

Additional Details : Weight : 8.02000kg

 View more versions of this product

Product Code. 15594046

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Manufactured from optically clear, virgin polystyrene
  • Treated surface helps cells adapt to reduced serum or serum-free media; stable at room temperature
  • Ideal for short- and long-term cell culture
  • Available with vented caps containing a 0.2μm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing risk of contamination; or phenolic-style caps for use in open systems requiring gas exchange
  • 3290 flask features redesigned neck for easier media pouring, lower height (1.4 in.) for reduced stacking height, and vented cap with Costar™-style concentric circle support grid geometry
  • Choice of canted or angled neck
    • Canted neck flasks allow easier pouring and improved access to the flask for pipetting or scraping
    • Angled neck improves pipetting access and reduces medium sloshing into the neck
  • Printed lot numbers provide easy traceability
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic
  • 100% integrity tested

  • Flasks 431328 and 431346 are prelabeled with bar code for use with SelecT™ Robotic System

Specifications

Specifications

No
HDPE
175cm2
PS (Polystyrene)
Yes
Angled
Sterile
35 to 52.5mL
Orange
Phenolic style
250mL
For optimal cell attachment
Lot numbers for ease in traceability
Rectangular
Corning™ Cellbind™
Videos