Learn More
Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Brand: Corning™ 354536
Code : 66
Additional Details : Weight : 0.34000kg
Description
Corning™ BioCoat™ products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.Specifications
2.36 oz. | |
70 mL | |
Canted | |
Rectangular | |
Sterile |
Phenolic | |
For adhesion, growth and differentiation of neuronal and transfected cell lines | |
10/cs. | |
Poly-D-Lysine | |
25 cm2 |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.