missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
Brand: Corning™ 354396
Code : 67
Additional Details : Weight : 0.50000kg
Description
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
Specifications
ANSI, SBS | |
With Lid | |
16 x 24 | |
Polystyrene | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
5/Cs. | |
Clear |
Standard | |
4°C to 30°C | |
Black, Clear Bottom | |
384 | |
Poly-D-Lysine | |
Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture | |
Poly-D-Lysine |